1H,1H,2H,2H-Nonafluorohexyl methacrylate stabilised
Leverandør: TCI
Synonymer: Methacrylic acid 3,3,4,4,5,5,6,6,6-Nonafluorohexyl ester, 3,3,4,4,5,5,6,6,6-Nonafluorohexyl methacrylate, Methacrylic acid 1H,1H,2H,2H-Nonafluorohexyl ester
Stabilisering: Stabilised with TBC
N1143-25G N1143-5G
TCIAN1143-25GEA 1725 NOK
TCIAN1143-25G TCIAN1143-5G
1H,1H,2H,2H-Nonafluorohexyl methacrylate stabilised
1H,1H,2H,2H-Nonafluorohexyl methacrylate
Formel: C₁₀H₉F₉O₂
MW: 332,17 g/mol
MDL nummer: MFCD00236094
CAS-nummer: 1799-84-4