Ramatroban

Leverandør: TCI

Synonymer: (R)-3-(3-(4-Fluorophenylsulphonamido)-3,4-dihydro-1H-carbazol-9(2H)-yl)propanoic acid

R0144-10MG R0144-50MG
TCIAR0144-10MGEA 1500 NOK
TCIAR0144-10MG TCIAR0144-50MG
Ramatroban
Ramatroban
Formel: C₂₁H₂₁FN₂O₄S
MW: 416,47 g/mol
MDL nummer: MFCD00887606
CAS-nummer: 116649-85-5

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR