2,2,6-Trimethylcyclohexanone

Leverandør: TCI

T3944-1G T3944-5G
TCIAT3944-1GEA 865 NOK
TCIAT3944-1G TCIAT3944-5G
2,2,6-Trimethylcyclohexanone
2,2,6-Trimethylcyclohexanone
Formel: C₉H₁₆O
MW: 140,23 g/mol
MDL nummer: MFCD00045550
CAS-nummer: 2408-37-9
FN: 1224

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR