N,N-Dietyl-p-fenylendiammoniumsulfat 97%
Leverandør: Thermo Scientific
Warning
Synonymer: N,N-Diethyl-p-phenylenediamine sulphate, N,N-Diethyl-1,4-phenylenediammonium sulphate, 4-Amino-N,N-diethylaniline sulphate, N1,N1-Diethylbenzene-1,4-diamine sulphate, N,N-Diethylbenzene-1,4-diammonium sulphate
A17832.0B A17832.18 A17832.30
A17832.0BEA 4700 NOK
A17832.0B A17832.18 A17832.30
N,N-Dietyl-p-fenylendiammoniumsulfat 97%
N,N-Dietyl-p-fenylendiammoniumsulfat
Purity: 97% CAS No.: 6283-63-2 Molecular Formula: C10H16N2.H2SO4 Formula Weight: 262.33 Melting Point: 183-188° Beilstein Reference: 4219768
Formel: C₁₀H₁₈N₂O₄S
MW: 262,33 g/mol
Kokepunkt: 274…275 °C (1013 hPa)
Smeltepunkt: 183…188 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00012993
CAS-nummer: 6283-63-2
EINECS: 228-500-6