1-Naftylamin ≥98%

Leverandør: Thermo Scientific
Danger

Synonymer: 1-Aminonaphthalene

104120250 104121000
104120250.EA 535 NOK
104120250. 104121000.
1-Naftylamin ≥98%
1-Naftylamin
Formel: C₁₀H₉N
MW: 143,19 g/mol
Kokepunkt: 301°C (760mmHg)
Flash punkt: 157°C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00004016
CAS-nummer: 134-32-7
EINECS: 205-138-7
FN: 2077
ADR: 6.1,III

Bestill nå

Spesifikasjon Testresultater

Appearance light yellow to light brown to dark purple crystalline powder
Infrared spectrum Conforms
GC ≥97.5 %
Melting point 48°C to 52°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR