2,4,6-Tri(dimetylaminometyl)fenol 90%

Leverandør: Thermo Scientific
Warning

Synonymer: Tris-2,6-dimethylaminomethylphenol, 2,6-Tris-N,N-dimethylaminomethylfenol, DMP-30, Epon® DMP-30 substitute

469290250 469291000
469290250.EA 685 NOK
469290250. 469291000.
2,4,6-Tri(dimetylaminometyl)fenol 90%
2,4,6-Tri(dimetylaminometyl)fenol
Formel: C₁₅H₂₇N₃O
MW: 265,4 g/mol
Kokepunkt: 130…135°C
Tetthet: 0,969
Flash punkt: 149°C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008330
CAS-nummer: 90-72-2
EINECS: 202-013-9
FN: 2735
ADR: 8,III

Bestill nå

Spesifikasjon Testresultater

Appearance colorless to yellow viscous liquid
Infrared spectrum Conforms
GC ≥88 %
Refractive index 1.5150 to 1.5170 (20°C, 589 nm)
Titration with HClO4 94 to 106 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR