Gallium (III) klorid, vannfri ≥98%

Leverandør: TCI

Synonymer: Gallium trichloride

G0359-25G G0359-5G
TCIAG0359-25GEA 2075 NOK
TCIAG0359-25G TCIAG0359-5G
Gallium (III) klorid, vannfri ≥98%
Gallium (III) klorid

CAS Number: 13450-90-3
MDL Number: MFCD00011018
Molecular Formula: Cl3Ga
Molecular Weight: 176.07
Form: Crystal
Color: White
Boiling point (°C): 201
Melting point (°C): 78

Formel: Cl₃Ga
MW: 176,08 g/mol
Kokepunkt: 201 °C
Smeltepunkt: 78 °C
MDL nummer: MFCD00011018
CAS-nummer: 13450-90-3
FN: 3260
ADR: 8,II

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR