Human IFN-gamma Quantikine ELISA Kit
Leverandør: Novus Biologicals
DIF50C PDIF50C SIF50C
NOVUDIF50CEA 0 NOK
NOVUDIF50C NOVUPDIF50C NOVUSIF50C
Human IFN-gamma Quantikine ELISA Kit
Assay ELISAs
The Quantikine Human IFN-gamma Immunoassay is a 4.5 hour solid phase ELISA designed to measure human IFN-gamma levels in cell culture supernates, serum, and plasma. It contains HEK293-expressed recombinant human IFN-gamma and antibodies raised against the recombinant factor.
Results obtained for naturally occurring human IFN-gamma samples showed linear curves that were parallel to the standard curves obtained using the Quantikine kit standards. These results indicate that this kit can be used to determine relative mass values for natural human IFN-gamma.
Store the unopened product at 2 to 8 °C.
New Product
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Primary antibody reactivity Human
Vert
Konjugert ELISA HRP