Millicell® Cell Culture Inserts, Mixed Cellulose Esters (MF-Millipore™) Membrane
Leverandør: Merck
PIHA01250 PIHA03050
MILFPIHA01250EA 5125 NOK
MILFPIHA01250 MILFPIHA03050
Millicell® Cell Culture Inserts, Mixed Cellulose Esters (MF-Millipore™) Membrane
Mikroplater Innsatsplater
With Millicell inserts, attachment or suspension cells can access media from both their apical and basolateral sides. Cell growth, structure, and function more closely mimic what occurs in vivo. In addition, Millicell inserts make it possible to study both sides of the cell monolayer.
The inserts are easily prepared for SEM and TEM visualising techniques, and they are compatible with cellular and/or fluorescent stains.
Bestill nå
SPESIFIKASJONER
Membranens effektive diameter 12 mm 30 mm
Environmentally Preferable
Total høyde 10,5 mm 13 mm