Pararosaniline hydrochloride
Leverandør: CHROMA
Danger
Synonymer: Basic Parafuchsin, 4,4'-(4-Iminocyclohexa-2,5-dienylidenemethylene)dianiline hydrochloride, [4-[Bis(4-aminophenyl)methylidene]-1-cyclohexa-2,5-dienylidene]azanium chloride, Basic Fuchsin hydrochloride, Parafuchsin hydrochloride, Pararosaniline chloride, Basic Red 9, 4,4'-((4-Iminocyclohexa-2,5-dien-1-ylidene)methylene)dianiline hydrochloride, Paramagenta hydrochloride
1B-0297.100 1B-0297.25 1B-297
CHRO1B-0297.100EA 0 NOK
CHRO1B-0297.100 CHRO1B-0297.25 CHRO1B-297
Pararosaniline hydrochloride
Pararosaniline hydrochloride
Formel: C₁₉H₁₈ClN₃
MW: 323,82 g/mol
Smeltepunkt: 268 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00001657
CAS-nummer: 569-61-9
EINECS: 209-321-2