VWR®, Overgang, reduksjon
Leverandør: VWR Collection
201-0052EA 220 NOK
201-0052 201-0053 201-0054 201-0055 201-0056 201-0057 201-0058 201-0059 201-0060 201-0061
VWR®, Overgang, reduksjon
Adaptere Adaptere med NS-slip
PTFE, tykke vegger.
  • Høy kjemikaliebestandighet
  • Gass-, væske- og vakuumtett
  • Temperaturbestandig fra −200...+280 °C
For greaseless connection of two different ground joint sizes; no grease residue can get into the analysis. Sturdy construction with grip and sealing rings, which prevent ground joints from seizing and give a very good seal.