Flaske/kanne, vid hals, med skrulokk, Nalgene®
Leverandør: Thermo Fisher Scientific
2226-0020 2226-0050
216-8731EA 2075 NOK
216-8731 216-8732
Flaske/kanne, vid hals, med skrulokk, Nalgene®
Flasker Kanner
PP, naturfarget, med hvit PP skrulokk, TPE pakning.
  • Very good chemical resistance, heat resistant up to 135 °C
  • Thick walled, heavy construction, rigid, robust, leakproof
  • Maintains a total vacuum for more than 8 hours at 20 °C in vacuum applications
Ideal for use as vacuum bottles. For vacuum applications, please use the quick filling and venting screw caps. Cat. Nos. 216-4385 to 216-4388. The bottles are not designed for use with overpressure. They are well suited as transfer and filtering bottles as well as for biotechnical applications.