Spruteflaske, vid hals, med trykt navn, DRIPLOK®, Azlon®
Leverandør: SciLabware
WGW001VTML WGW020VTML WGW002VTML WGW004VTML
215-3151EA 800 NOK
215-3151 215-3279 215-3278 215-3777
Spruteflaske, vid hals, med trykt navn, DRIPLOK®, Azlon®
Flasker Spruteflasker
LDPE, gjennomskinnelig med PP sprayhette.
  • Wide mouth for easy filling
  • Supplied with adhesive hazard labels
Do-it-yourself DripLok® wash bottles with blank write-on panel.
Sertifiseringer: Azlon® wash bottles meet the material requirements of the U.S. Food and Drug Administration as specified in 21 CFR 177.1520.
Bestillingsinformasjon: Assorted colour version with each one orange, blue, green, yellow and red cap.