Stainless Steel Buckets
Leverandør: RSG
730.006 730.010 730.015 730.012
216-1520EA 1625 NOK
216-1520 216-1540 216-1541 216-0271
Stainless Steel Buckets
Bøtter
Stainless steel, rust-proof, polished finish.
  • Very good chemical and thermal resistance
  • High quality, very stable
  • Optional matching lids are available
For use in laboratories and industry. Graduated with handle. Seamless handle tabs and smooth surfaces for fast, easy cleaning.