Inoculating loops, disposable, Microloops
Leverandør: MEDICAL WIRE EQUIPMENT
MW700/1 MW701/1 MW710/20
300-0500EA 2850 NOK
300-0500 300-0511 300-0510
Inoculating loops, disposable, Microloops
Utstryksspatler Podeøser og Podenåler
Sterile, disposable inoculating loops.
  • Colour coded plastic
  • Moderately flexible