Inoculating loops, metal
Leverandør: Copan
310C25T 311C25T 312C25T 313C25T
612-2668EA 2850 NOK
612-2668 612-2669 612-2670 612-2671
Inoculating loops, metal
Utstryksspatler Podeøser og Podenåler
These nickel-chrome metal wire loops are available non calibrated in a range of sizes with internal diameters (Ø) of 2, 3, 4 or 5 mm, or calibrated, 1 and 10 μl sizes, for performing quantitative specimen cultures, such as urinalysis.
  • Loops have totally smooth surfaces and will not cut or gouge through agar medium when streaking plate cultures
  • Durable 80/20 nickel-chrome with flexible twisted metal stem for flexibility and rapid cooling after sterilisation
  • Designed to be durable and long lasting, whilst being more cost effective than platinum-rhodium
Forpakning: Non calibrated loops are supplied in tubes containing 25 loops; calibrated loops are supplied individually in tubes with a Certificate of Calibration.