Inokuleringsløkker, gjenbrukbare
Leverandør: RSG
268.001 268.003 268.002 268.013 268.012 268.011
631-7131EA 415 NOK
631-7131 631-7133 631-7132 631-0086 631-0085 631-0084
Inokuleringsløkker, gjenbrukbare
Utstryksspatler Podeøser og Podenåler
These NiCr 8020 loops are available in a range of sizes and lengths with internal diameter of 1,5, 2,5 or 4 mm. They are heat resistant and can be flame sterilised. Diameter of wire: 0,6 mm. Loop holder available separately.