VWR® Protective Covers for Clamp Prongs
Leverandør: VWR Collection
241-0152EA 750 NOK
241-0152 241-0153
VWR® Protective Covers for Clamp Prongs
Deksler
These protecting covers for clamp jaws are made from red vinyl.
  • Good chemical and thermal stability
  • Fits snugly but easily over the clamp jaws
  • Protects clamped items
Suitable for VWR® clamps with prongs.