Skrulokk, rask påfylling/-ventilasjon, med overganger, Nalgene®
Leverandør: Thermo Fisher Scientific
2158-0022 2158-0021 2158-0031 2158-0032
216-4386EA 5425 NOK
216-4386 216-4385 216-4387 216-4388
Skrulokk, rask påfylling/-ventilasjon, med overganger, Nalgene®
Lokk Flaskelokk
PP, hvit, TPE-pakning.
  • Very good chemical resistance, heat resistant from −10 to +135 °C, autoclavable
  • Available with two or three hose connectors for inner hose diameter of 6 or 9 mm
  • With safety shut-off valve
Screw closure (closure size 83B) with quick-release closures and inner and outer tubing connectors. For a convenient transfer of liquids with customer's own tubing. Ideal for use as a vacuum trap with Nalgene® carboys for high loads. Each connection on the lid has a shut-off valve that closes when a hose connector is removed.