Cation exchange HPLC columns, PRP-X200
Leverandør: HAMILTON BONADUZ
79394 79441 79442 79384 79357 79456 79368 79449 79369 79127 79128 79363 79288 79914
554-0223EA 11625 NOK
554-0223 554-0243 554-0244 554-0216 554-0207 554-1270 554-1245 554-1264 554-1246 554-1227 554-1228 554-1493 HAMI79288 HAMI79914
Cation exchange HPLC columns, PRP-X200
Kromatografikolonner
Polymeric cation exchange packing for separation of inorganic and organic cations.
  • pH stable from 1 to 13
  • Easily seperates mono or divalent cations, alcalins et alcalino-terreux
  • Use with organic solvent from 0 to 100% for elution of hydrophobic cations and column cleaning
  • Use conductivity or UV detection