Anion exchange HPLC columns, PRP-X600
Leverandør: HAMILTON BONADUZ
79189 79361 79362 79360 79220
554-1239EA 19750 NOK
554-1239 554-1243 554-1244 554-0208 HAMI79220
Anion exchange HPLC columns, PRP-X600
Kromatografikolonner
PRP-X600 is a superficially porous weak-base anion exchange support that separates DNA oligomers according to negative charge.
  • Fast separation with better sample capacity than non-porous supports
  • Improved sample recovery due to minimised hydrophobic interactions
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Maks. trykk 5000 psi
pH område 1 - 13
Temperaturgrense 5 - 60 °C (pH 1 - 7,9); 5 - 30 °C (pH 8 - 13)