Spiralkjøler med NS-slip
Leverandør: Lenz Laborglas GmbH & CO.KG
05233404 05233504 05233201
201-3353EA 3975 NOK
201-3353 LENZ05233504 271-3351
Spiralkjøler med NS-slip
Kondensator
DURAN®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
For cooling the condensate. Water supply via glass tubing connectors with 11 mm external Ø.