Prøvebeholder, med skrulokk
Leverandør: KARTELL
916 914 910 912 913 911
216-3112EA 2300 NOK
216-3112 216-3110 216-3102 216-3106 216-3108 216-3104
Prøvebeholder, med skrulokk
Prøvecontainere Beholdere, generelt bruk
To store and transport samples securely.
LDPE, translucent.
  • Good chemical resistance, good temperature resistance
  • Strong walls, slightly conical shape
  • Particularly tight seal through screw cap with cone gasket