VWR®, Conical Sample Containers, with Snap-On Lids
Leverandør: VWR Collection
216-2639EA 6625 NOK
216-2639 216-2640 216-2648 216-2651 216-2652 216-2649
VWR®, Conical Sample Containers, with Snap-On Lids
Prøvecontainere Beholdere, generelt bruk
PP, translucent.
  • Available in sterile or non sterile versions
  • Raised graduations
  • Leakproof snap-on lid
Conical containers for sample collection, storage and transport.
NB: *The product(s) on this page are outside the scope of the EU Directive 2019/904.