Retter, fordamper
Leverandør: MORGAN ADVANCED MATERIALS HALD
L131/2 L131/3 L131/4 L131/5 L131/6 L131/8
216-0370EA 405 NOK
216-0370 216-0371 216-0289 216-0372 216-0373 216-0375
Retter, fordamper
Skåler Fordampingsskåler
Porselen, glasert.
  • Høy kjemikaliebestandighet
  • Varmebestandig opptil +1000 °C
  • Lav varmeutvidelse
For a wide range of laboratory applications. With spout and flat bottom. White glaze inside and outside.