Filterpapir, frøkontroll, Whatman™
Leverandør: Whatman products (Cytiva)
10344672 10344676 10345572 10345576 10342580 10342583 10342579 10342581 10342555 10342570 10345573
515-8115EA 37750 NOK
515-8115 512-1165 512-1166 512-1167 515-5014 515-5015 512-0647 WHAT10342581 WHAT10342555 WHAT10342570 WHAT10345573
Filterpapir, frøkontroll, Whatman™
Filter Filterpapir Kvalitativt filterpapir
Owing to their extremely high purity our seed testing papers provide reliable and reproducible results. The papers are made from pure cellulose without any additives and do not contain any substances that could influence the growth of the seeds. The constant water absorption of the papers ensures the continuous provision of the required amount of water.
Grade 3645 sheets are suitable for testing seeds with small white rootlets.
Grade 3014 sheets are suitable for testing particulalry sensitive seeds. Medium-large and coated seeds (e.g. sugar beet, fodder beet, grain, sunflower, rapeseed,More Product Information
Bestill nå
SPESIFIKASJONER
Nr. 3014 3014 3236 3644 3645 3621
Environmentally Preferable
Vekt 113 g/m² 110 g/m² 720 g/m² 165 g/m² 700 g/m²
Tykkelse 0,22 mm 0,22 mm 0,22 mm 1,40 mm 0,35 mm -