Glassmikrofiberfiltre med bindemiddel, grade GF 10, Whatman™
Leverandør: Whatman products (Cytiva)
10370302 10370305 10370308 10370319 10370320 10370394 10370391 10370393 10370395 10370381 10370384 10370392 10370429 10370435
513-1000EA 2500 NOK
513-1000 513-1001 513-1002 513-1003 513-1004 513-0089 512-0663 513-0121 WHAT10370395 512-0662 513-0119 513-0120 WHAT10370429 WHAT10370435
Glassmikrofiberfiltre med bindemiddel, grade GF 10, Whatman™
Filter Glassfiberfilter
These glass microfibre filters made from borosilicate glass are strengthened with an organic binder to provide high mechanical stability.
  • Medium filtration speed
  • Can be used as a roll filter in automated applications
These filters can be used as a weighing aid during infrared weighing or for automatic sampling applications. They also come in reel format for use as a roll filter in automatic air filtration units.
Bestill nå
SPESIFIKASJONER
Nr. GF 10
Environmentally Preferable
Vekt 70 g/m² 70 g/m²
Tykkelse 350 µm 350 µm
Luftgjennomstrømning 12 s/100 ml/1,56 cm²
Maks. temp omgivelser 180 °C 180 °C