Sprøytefilter, Minisart® NY
Leverandør: Sartorius
17845-ACK 17845-Q 17846-ACK 17846-Q 1784B-K 1784B-Q 1784C-Q 1784C-K 1776B-K 1776B-Q 1776C-K 1776C-Q 17846-R 17845-R
514-0080EA 3225 NOK
514-0080 514-0081 514-0082 514-0083 SART1784B-K SART1784B-Q SART1784C-Q 516-0684 SART1776B-K SART1776B-Q 516-0681 SART1776C-Q SART17846-R SART17845-R
Sprøytefilter, Minisart® NY
Filter Sprøytefilter
Minisart® NY with hydrophilic polyamide (nylon) is optimised for aqueous liquids and solvents with a pH of 3 to 14. Type 17845 with a pore size of 0,2 µm can be used for sample preparation and sterile filtration. It has a male Luer slip outlet and version ACK is individually packed and sterilised by ethylene oxide (EO).
  • Sterile and ready-to-use
  • Minimum extractables or leachables
  • Low non-specific binding
  • Outperforming chemical compatibility
Individually packed and sterile Minisart® NY syringe filters are optimal for immediate usage whenever sterileMore Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Porestørrelse 0,2 µm 0,45 µm 0,2 µm 0,45 µm 0,2 µm
Luftgjennomstrømning 50 ml/min with water at 1 bar (14,5 psi) | 70 ml/min with methanol at 1 bar (14,5 psi) 100 ml/min with water at 1 bar (14,5 psi) | 200 ml/min with methanol at 1 bar (14,5 psi) 20 ml/min with water at 1 bar (14,5 psi) | 40 ml/min with methanol at 1 bar (14,5 psi) 40 ml/min with water at 1 bar (14,5 psi) | 110 ml/min with methanol at 1 bar (14,5 psi) 100 ml/min with water at 1 bar (14,5 psi) | 200 ml/min with methanol at 1 bar (14,5 psi)
Maks. bemanningstrykk 4,5 bar (65 psi) 4,5 bar (65 psi) 4,5 bar (65 psi) 4,5 bar (65 psi) 4,5 bar (65 psi)
Maks. temp omgivelser 121 °C (250 °F) 121 °C (250 °F) 121 °C (250 °F) 121 °C (250 °F) 121 °C (250 °F)