Cellekulturflaske, Falcon®
Leverandør: Corning
353014 353018 353024 353028 353082 353108 353109 353112 353009 353133 353107 353110 353135 353136 353138 353139 355000 355001
734-0009EA 3000 NOK
734-0009 734-0010 734-0012 734-0014 734-0031 734-0044 734-0045 734-0047 734-0962 734-0965 734-0043 734-0046 734-0049 734-0050 734-0957 734-1031 734-0266 734-0267
Cellekulturflaske, Falcon®
Kolber Celledyrkningsflasker
PS, cellekulturbehandlede, sterile, pyrogenfrie.
  • Easy to read printed volumetric graduations, and writing patch
  • Vented caps incorporate a 0,2 µm hydrophobic membrane
  • Convenient, peel-open, medical-style packaging ensures flask sterility is maintained
Vacuum gas plasma tissue culture treatment ensures consistent cell attachment, spreading and growth.
Barcoded (robotics/automation compatible).
NB: Merk: verdiene for dyrkingsområdet og kapasiteten er nominelle.