Cell culture flasks, double side arm Celstir®, with heating jacket, WHEATON®
Leverandør: DWK Life Sciences
356949 356943 356945 356946 356952 356954
734-3181EA 25225 NOK
734-3181 734-3178 734-3179 734-3180 734-3182 734-3183
Cell culture flasks, double side arm Celstir®, with heating jacket, WHEATON®
Kolber Spinnerflaske
Borosilicate glass flask, PTFE and glass impeller assembly, PTFE and silicone lined top cap.
Double side arm Celstir® flasks complete with water jacket allows precise temperature control of flask contents when operated with a recirculating water bath. Hose connectors accept 6 mm tubing.