Volumetric flasks, class A, blue markings
Leverandør: HIRSCHMANN
2820266 2820261 2820275 2820278 2820281
612-5738EA 865 NOK
612-5738 HIRS2820261 HIRS2820275 HIRS2820278 HIRS2820281
Volumetric flasks, class A, blue markings
Kolber Målekolber
DURAN®, borosilicate glass, with glass stopper, conformity certified (DE-M marking).
  • Blue enamelled indelible markings resistant to acids and alkalis
  • Certificate of conformity with dated batch identification (available via website: www.hirschmannlab.de)
* In addition to DIN
**Wide neck
Sertifiseringer: DIN EN ISO 1042