Målekolbe, klasse A, blå gradering, BLAUBRAND®
Leverandør: Brand
37045 37053 37046 37047 37048 37049 37050 37051 37052
612-2736EA 570 NOK
612-2736 612-3565 612-2563 612-2564 612-2565 612-2566 612-2567 612-2568 612-2569
Målekolbe, klasse A, blå gradering, BLAUBRAND®
Kolber Målekolber
Borosilikatglass 3.3, uten propp.
  • Calibrated to contain (TC, In)
  • Marks and inscriptions in high contrast blue enamel
  • DE-M marking
* Wide neck.
Sertifiseringer: DIN EN ISO 1042
Bestillingsinformasjon: Supplied with a batch certificate. Volumetric flasks also available on request with an individual certificate, a USP individual certificate or a DAkkS calibration certificate.