Trakter, analytisk med lang stilk, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
F146440000 F146480000 F146500000 F146400000 F146420000
221-0279EA 1200 NOK
221-0279 221-1243 221-1244 221-1245 221-1246
Trakter, analytisk med lang stilk, SP Bel-Art
Trakter Filtreringstrakter Analysetrakter
PMP, gjennomsiktig.
  • Good chemical resistance
  • Heat resistant to 177 °C
  • Vertical outer ribs prevent airlock
Urbanti type funnel with internal helicoid ribs for fast filtration. This produces a whirlpool effect so that liquids flow through many times faster. This effect is achieved even with standard filter paper inserted in the funnel. Air is prevented from being trapped between the filter paper and the funnel.