Filter funnels acc. to Pregl, VitraPOR®
Leverandør: ROBU GLASFILTER
28040 28041 28043 28044 28042
ROBU28040EA 4225 NOK
ROBU28040 ROBU28041 ROBU28043 ROBU28044 ROBU28042
Filter funnels acc. to Pregl, VitraPOR®
Trakter Filtreringstrakter Mikrofiltertrakter
Borsilikatglass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Med innebygd, sintret glassfilterskive.