Skilletrakt, konisk, Squibb, Nalgene®
Leverandør: Thermo Fisher Scientific
4301-0125 4301-0250 4301-0500 4301-1000 4301-2000
527-2101EA 3450 NOK
527-2101 527-2102 527-2103 527-2104 527-2105
Skilletrakt, konisk, Squibb, Nalgene®
Trakter Skilletrakter
FEP translucent, TFE stopcock, ETFE screw closure.
  • Leakproof closure and stopcock
  • Autoclavable except for stopcock assembly
  • TFE stopcock requires no lubrication
Robust, transparent, non stick for easy cleaning, non wetting for complete draining and impervious to any chemical used in a separating funnel.