Glassfilter, dypperør, VitraPOR®
Leverandør: ROBU GLASFILTER
25100 25101 25102 25103 25104
ROBU25100EA 2825 NOK
ROBU25100 ROBU25101 ROBU25102 ROBU25103 ROBU25104
Glassfilter, dypperør, VitraPOR®
Filter Filterrør
Micro immersion filter suitable for reverse filtration.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion
Made of borosilicate glass 3.3.