VWR®, Colour-Frosted Microscope Slides
Leverandør: VWR
VWR®
631-1564EA 160 NOK
631-1564 631-1563 631-1562 631-1561 631-1557 631-1556 631-1555 631-1565 631-1554 631-1559 631-1558 631-1560
VWR®, Colour-Frosted Microscope Slides
Objektglass Objektglass til mikroskop
Mattede ender i seks forskjellige farger.
  • Available with either cut or bevelled edges
  • Writing surface resistant to all common dyes and solvents