VWR®, Cover Slips, Rectangular
Leverandør: VWR Collection
631-1569EA 2425 NOK
631-1569 631-1572 631-1573 631-1574 631-1575
VWR®, Cover Slips, Rectangular
Dekkglass
Dekkglassene er egnet for diagnostikk.
  • Made of pure white Hydrolytic Class 1 glass
Thickness: 0.13 - 0.16 mm
Thickness: 0,13 - 0,16 mm
Forpakning: Pakket i dispenseresker i plast.