Filterspisser, Solvent Safe™
Leverandør: Thermo Fisher Scientific
5079EB 5479E
612-9621EA 0 NOK
612-9621 612-9643
Filterspisser, Solvent Safe™
Pipettespisser
Karbonfilteret Solvent Safe™ beskytter pipettene mot skade ved å blokkere korrosive damper fra flyktige organiske løsningsmidler og er et billigere alternativ til dyre, positive displacement pipetter.
  • The Solvent Safe™ carbon filter protects pipettes from damage by blocking corrosive vapours from volatile organic solvents and is a less costly alternative to expensive positive displacement pipettes