Robotspiss for Qiagen® BioRobot® (3000/8000/9600/9604, Rosys, Colibri)
Leverandør: Thermo Fisher Scientific
953-251 953-252 953-011 951-251 951-011 951-021 951-252 951-253 951-261 951-262 953-021 953-253 953-261 953-262
737-2028EA 3525 NOK
737-2028 737-2030 736-0032 737-2020 736-0031 737-2019 737-2022 737-2024 737-2025 737-2026 736-0033 737-2032 737-2033 737-2034
Robotspiss for Qiagen® BioRobot® (3000/8000/9600/9604, Rosys, Colibri)
Pipettespisser
Spiss av høy kvalitet. Den er beregnet for å sikre kompatibiliteten og en bra passform.
  • PP, klar eller sort
  • Spiss med ART®-filter: eliminerer risikoen for krysskontaminering, utformet for bruk med DNA, RNA, radioaktive og infeksiøse prøver
  • Garantert RNase, DNase, ATP og pyrogenfri
  • Steril spiss: Gammabestrålt
Bestillingsinformasjon: Fås i forpakninger med 10 eller 50 stativer (Econopak).
Bestill nå
SPESIFIKASJONER
Environmentally Preferable Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact
Miljøsertifikat IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603) IRECS; ISO 14001 (Certificate Registration No. EMS 752603)