Racks for centrifuge tubes, Poxygrid®, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
F187940000 F187940001 F187940002
525-9101EA 1800 NOK
525-9101 525-9102 525-9134
Racks for centrifuge tubes, Poxygrid®, SP Bel-Art
Stativer for rør Tube Racks
Epoxy coated steel wire, autoclavable.
  • Ideal for supporting conical bottom centrifuge tubes
  • The bottom rails hold the tip of the conical tube
Hole Ø: 30 mm, for 525-9100: 18 mm