Racks for hydrometers, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
F189880001 F189880000
614-0001EA 4000 NOK
614-0001 614-0000
Racks for hydrometers, SP Bel-Art
Stativer for rør Aerometerstativer
Polypropylene racks, both designed to hold up to 18 hydrometers. Top plate and middle plate have 25 mm diameter holes, bottom plate has wells tapering from 25 to 9 mm to accept hydrometer bulb.
Overall height of both racks is 250 mm.