Safety spectacles, uvex skylite
Leverandør: UVEX
9174.065 9174.066 9174.095
111-8262EA 345 NOK
111-8262 111-8261 111-8263
Safety spectacles, uvex skylite
Vernebriller
Single lens safety glasses with clear or grey lens offering best protection from UV radiation.
  • Frosted anti-dazzle side shields
  • Adjustable lens angle
  • Anti-scratch coating
Weight: 26 g
Sertifiseringer: EN 166-168, EN 170/172; 1F