Svaber, med transportmedium for virus og klamydia
Leverandør: Copan
144C 147C 148C 145C
710-0450EA 940 NOK
710-0450 710-0453 710-0454 710-0451
Svaber, med transportmedium for virus og klamydia
Svabere Svabere for transport
Liquid transport medium is contained in a sponge carrier located in the bottom of the tube.
  • Special colours of caps to make differentiation easy
  • Simple insert swab into the tube after collecting sample
  • Swab tip placed in direct contact with the medium soaked sponge
Sertifiseringer: ISO 13485. ISO 9001:2008.
Forpakning: Individually sealed in a peel-pouch. 25 swabs per box, 10x25 swabs per case.
NB: *Produktet/produktene på denne siden er utenfor omfanget av EU -direktivet 2019/904