Microsyringes for hewlett-packard systems
Supplier: Hospidex
EX-MSEX10HP26 EX-MS10HP23/26
613-1736EA 1150 NOK
613-1736 613-1740
Microsyringes for hewlett-packard systems
Syringes Syringes for Chromatography
Microsyringes for Agilent instruments.
Type 1: Ti-Ni alloy elastic piston.
Type 2: Stainless steel piston.
Order Now