Tapedispenser
Leverandør: PRECISION DYNAMICS TIMEM
500P 60P HD-2
817-1592EA 2125 NOK
817-1592 817-1645 817-0312
Tapedispenser
Etikett- og tapedispensere
Plastdispensere for flere taperuller. Har flipplokk og sugekopper under.
  • Metaakrylat
For ruller med en bredde på 13 eller 55 mm.
60P: Bench top dispenser for rolls with 76 mm cores. Can hold multiple rolls of tape and label products up to a total of 235 mm. Size: 250×184×152 mm.
500P: Bench top dispenser for rolls with 25 mm cores. Can hold multiple rolls of tape and label products up to a total of 228 mm. Size: 250×114×86 mm.