VWR®, lommetermometer
Leverandør: VWR Collection
620-0809EA 480 NOK
620-0809 620-0810 620-0853
VWR®, lommetermometer
Termometere Analogue Thermometers
These glass thermometers have a PTFE coating that helps to prevent contamination and contains liquid and glass in the event of breakage. Red spirit. Total immersion thermometers. Ideal for field studies, wastewater treatment and any 'on the go' applications.
  • Bulbous glass body
  • Individual serial number
Accuracy: ±1,0 °C