Triangler for digler
Leverandør: USBECK Laborgeräte
5100 5101 5102 5104
451-9301EA 81 NOK
451-9301 451-9302 451-9303 451-9387
Triangler for digler
Trefot
Triangles made from galvanised steel wire, with clay tubes.