Mikrosentrifugerør, 0,5/1,5/2,0 ml
Leverandør: Brand
780730 780731 780732 780733 780734 780740 780741 780742 780743 780744 780750 780751 780752 780753 780754
211-3231EA 2425 NOK
211-3231 211-3232 211-3233 211-3234 211-3235 211-3241 211-3242 211-3243 211-3244 211-3245 211-3236 211-3237 211-3238 211-3239 211-3240
Mikrosentrifugerør, 0,5/1,5/2,0 ml
Rør Mikrosentrifugerør
PP, graderte, sterile med PP-skrulokk eller ikke-sterile uten PP-skrulokk, autoklaverbare.
  • Can withstand centrifugation to 17000×g
  • Cap with silicone seal
  • With frosted marking area
  • Sterile tubes are DNA, DNase, and RNase-free, endotoxin-free, non-mutagenic, non-toxic
Temperature range: −196 to +121°C.
*without graduations.