Test tubes with rim, DURAN®
Leverandør: DWK Life Sciences
261100109 261100306 261100606 261100803 261101105 261101302 261101602 261102607 261102804 261103303 261103603 261103809 261102101 261102307
212-1202EA 485 NOK
212-1202 212-1204 212-1206 212-1208 212-1210 212-1212 212-1214 212-1220 212-1222 212-1224 212-1226 212-1228 212-1216 212-1218
Test tubes with rim, DURAN®
Rør Reagensrør
FIOLAX®, neutral glass.
  • Excellent chemical resistance
  • Relatively resistant against rapid temperature changes and local heating
Thin walled test tubes for a wide variety of applications.